missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Carbamoylphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
$147.25 - $147.25
Specifications
| CAS | 123088-59-5 |
|---|---|
| Melting Point | 234°C |
| Molecular Formula | C7H8BNO3 |
| MDL Number | MFCD03411940 |
| InChI Key | GNRHNKBJNUVWFZ-UHFFFAOYSA-N |
Specifications
| 123088-59-5 | |
| C7H8BNO3 | |
| GNRHNKBJNUVWFZ-UHFFFAOYSA-N | |
| 2737811 | |
| Crystalline Powder |
| 234°C | |
| MFCD03411940 | |
| (4-carbamoylphenyl)boronic acid | |
| 164.96 |