accessibility menu, dialog, popup

UserName

CAS RN 123088-59-5

IUPAC Name: (4-carbamoylphenyl)boronic acid
Synonyms: 4-carbamoylphenylboronic acid
Molecular Weight (g/mol): 164.96
Molecular Formula: C7H8BNO3
InChi Key: GNRHNKBJNUVWFZ-UHFFFAOYSA-N
SMILES: NC(=O)C1=CC=C(C=C1)B(O)O
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results