missing translation for 'onlineSavingsMsg'
Learn More

4-Butylphenyl Isocyanate 98.0+%, TCI America™

Catalog No. p-7122807
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B32051G 1 g
B32055G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B32051G Supplier TCI America Supplier No. B32051G
Only null left

Chemical Identifiers

CAS 69342-47-8
Molecular Formula C11H13NO
Molecular Weight (g/mol) 175.231
MDL Number MFCD00013882
InChI Key LJJRXPXDTAUVQU-UHFFFAOYSA-N
PubChem CID 2796313
IUPAC Name 1-butyl-4-isocyanatobenzene
SMILES CCCCC1=CC=C(C=C1)N=C=O

Specifications

Color Yellow
Boiling Point 170°C
Quantity 1 g
UN Number 2206
Synonym 4-butylphenyl isocyanate, 4-n-butylphenyl isocyanate, benzene, 1-butyl-4-isocyanato, benzene,1-butyl-4-isocyanato, 4-butylbenzenisocyanate, 4-butylphenylisocyanate, acmc-209o7f, 1-butyl-4-isocyanato-benzene, 4-n-butyl phenyl isocyanate
Formula Weight 175.23
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Butylphenyl Isocyanate