missing translation for 'onlineSavingsMsg'
Learn More

4-Butylphenol 96.0+%, TCI America™

Catalog No. p-7118812
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B10265G 5 g
B102625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B10265G Supplier TCI America Supplier No. B10265G
Only null left

Chemical Identifiers

CAS 1638-22-8
Molecular Formula C10H14O
Molecular Weight (g/mol) 150.221
MDL Number MFCD00041750
InChI Key CYYZDBDROVLTJU-UHFFFAOYSA-N
Synonym 4-n-butylphenol, p-butylphenol, phenol, 4-butyl, p-hydroxybutylbenzene, phenol, p-butyl, 1-hydroxy-4-n-butylbenzene, p-n-butylphenol, 4-butyl-phenol, 4-n-ethylphenol
PubChem CID 15420
ChEBI CHEBI:34437
IUPAC Name 4-butylphenol
SMILES CCCCC1=CC=C(C=C1)O

Specifications

Color White-Yellow
Boiling Point 245°C
Quantity 5 g
UN Number 2430
Formula Weight 150.22
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 4-Butylphenol
EINECSNumber (3)-0503&(4)-0057
RTECSNumber SJ8922500
TSCA Yes