missing translation for 'onlineSavingsMsg'
Learn More

4-Butylmorpholine 98.0+%, TCI America™

Supplier:  TCI America B46025ML

Catalog No. B46025ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

4-Butylmorpholine
Colorless
C8H17NO
5 mL
LMRKVKPRHROQRR-UHFFFAOYSA-N
4-butylmorpholine
70499
≥98.0% (GC,T)
1005-67-0
78°C
MFCD00023401
1993
CCCCN1CCOCC1
143.23
143.23
Liquid

Chemical Identifiers

1005-67-0
143.23
LMRKVKPRHROQRR-UHFFFAOYSA-N
4-butylmorpholine
C8H17NO
MFCD00023401
70499
CCCCN1CCOCC1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (5)-0860

RTECSNumber : QD8773000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions