missing translation for 'onlineSavingsMsg'
Learn More

4-Butylcyclohexanol (cis- and trans- mixture) 98.0+%, TCI America™

Catalog No. b19305g
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B19305G 5 g
B193025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B19305G Supplier TCI America Supplier No. B19305G
Only null left

Chemical Identifiers

CAS 70568-60-4
Molecular Formula C10H20O
Molecular Weight (g/mol) 156.269
MDL Number MFCD01321147
InChI Key NUANGSLQWFBVEH-UHFFFAOYSA-N
PubChem CID 4132186
IUPAC Name 4-butylcyclohexan-1-ol
SMILES CCCCC1CCC(CC1)O

Specifications

Color Colorless
Boiling Point 122°C
Quantity 5 g
Formula Weight 156.27
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Butylcyclohexanol (cis- and trans- mixture)
EINECSNumber (3)-2301
TSCA No