missing translation for 'onlineSavingsMsg'
Learn More

4-Butylbenzyl Bromide 97.0+%, TCI America™

Catalog No. p-7119888
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B30051G 1 g
B30055G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B30051G Supplier TCI America Supplier No. B30051G

Chemical Identifiers

CAS 10531-16-5
Molecular Formula C11H15Br
Molecular Weight (g/mol) 227.145
InChI Key XNQBGVFFKGZXJH-UHFFFAOYSA-N
PubChem CID 12150840
IUPAC Name 1-(bromomethyl)-4-butylbenzene
SMILES CCCCC1=CC=C(C=C1)CBr

Specifications

Color Yellow
Boiling Point 144°C
Quantity 1 g
UN Number 3265
Formula Weight 227.15
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Butylbenzyl Bromide