missing translation for 'onlineSavingsMsg'
Learn More

4-Butylbenzene-1-sulfonyl chloride, 97%, Thermo Scientific™

Chemical Identifiers

CAS 54997-92-1
Molecular Formula C10H13ClO2S
Molecular Weight (g/mol) 232.72
MDL Number MFCD00173908
InChI Key OVFZELSNOHIDEF-UHFFFAOYSA-N
Synonym 4-butylbenzene-1-sulfonyl chloride, 4-n-butylbenzenesulfonyl chloride, 4-n-butyl benzenesulphonyl chloride, 4-butyl-benzenesulfonyl chloride, 4-n-butylphenylsulfonyl chloride, 4-n-butylbenzenesulphonyl chloride, benzenesulfonyl chloride, 4-butyl, 4-butylphenyl chlorosulfone, pubchem5484, 4-n-butylbenzenesulfonylchloride
PubChem CID 2737352
IUPAC Name 4-butylbenzene-1-sulfonyl chloride
SMILES CCCCC1=CC=C(C=C1)S(Cl)(=O)=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
BTB10243EB
View Documents
N/A 25g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

54997-92-1
232.72
OVFZELSNOHIDEF-UHFFFAOYSA-N
2737352
CCCCC1=CC=C(C=C1)S(Cl)(=O)=O
C10H13ClO2S
MFCD00173908
4-butylbenzene-1-sulfonyl chloride, 4-n-butylbenzenesulfonyl chloride, 4-n-butyl benzenesulphonyl chloride, 4-butyl-benzenesulfonyl chloride, 4-n-butylphenylsulfonyl chloride, 4-n-butylbenzenesulphonyl chloride, benzenesulfonyl chloride, 4-butyl, 4-butylphenyl chlorosulfone, pubchem5484, 4-n-butylbenzenesulfonylchloride
4-butylbenzene-1-sulfonyl chloride

Specifications

54997-92-1
100.0
C10H13ClO2S
MFCD00173908
OVFZELSNOHIDEF-UHFFFAOYSA-N
4-butylbenzene-1-sulfonyl chloride
2737352
97%
96.5
97%
CH3(CH2)3C6H4SO2Cl
4-butylbenzene-1-sulfonyl chloride, 4-n-butylbenzenesulfonyl chloride, 4-n-butyl benzenesulphonyl chloride, 4-butyl-benzenesulfonyl chloride, 4-n-butylphenylsulfonyl chloride, 4-n-butylbenzenesulphonyl chloride, benzenesulfonyl chloride, 4-butyl, 4-butylphenyl chlorosulfone, pubchem5484, 4-n-butylbenzenesulfonylchloride
CCCCC1=CC=C(C=C1)S(Cl)(=O)=O
232.72
232.73
4-Butylbenzene-1-sulfonyl chloride
Videos
Safety and Handling

Safety and Handling

GHS P Statement Causes severe skin burns and eye damage. Contact with water liberates toxic gas. Reacts violently with water.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Immediately call a POISON CENTER or doctor/physician. IF IN EYES: Rinse cau

Danger

SDS
Documents

Documents