missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Butylaniline 97.0+%, TCI America™
$68.97 - $445.51
Chemical Identifiers
| CAS | 104-13-2 |
|---|---|
| Molecular Formula | C10H15N |
| Molecular Weight (g/mol) | 149.237 |
| MDL Number | MFCD00007925 |
| InChI Key | OGIQUQKNJJTLSZ-UHFFFAOYSA-N |
| Synonym | 4-n-butylaniline, benzenamine, 4-butyl, p-n-butylaniline, p-butylaniline, 4-butylbenzenamine, p-aminobutylbenzene, p-butylaminobenzene, 1-amino-4-butylbenzene, p-butyl aniline, 4-butylphenylamine |
| PubChem CID | 7694 |
| IUPAC Name | 4-butylaniline |
| SMILES | CCCCC1=CC=C(C=C1)N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
B027225ML
|
TCI America
B027225ML |
25 mL |
Each for $68.97
|
|
|||||
|
B0272100ML
|
TCI America
B0272100ML |
100 mL |
Each for $134.21
|
|
|||||
|
B0272500ML
|
TCI America
B0272500ML |
500 mL |
Each for $445.51
|
|
|||||
Chemical Identifiers
| 104-13-2 | |
| 149.237 | |
| OGIQUQKNJJTLSZ-UHFFFAOYSA-N | |
| 7694 | |
| CCCCC1=CC=C(C=C1)N |
| C10H15N | |
| MFCD00007925 | |
| 4-n-butylaniline, benzenamine, 4-butyl, p-n-butylaniline, p-butylaniline, 4-butylbenzenamine, p-aminobutylbenzene, p-butylaminobenzene, 1-amino-4-butylbenzene, p-butyl aniline, 4-butylphenylamine | |
| 4-butylaniline |
Specifications
| 104-13-2 | |
| C10H15N | |
| 25 mL | |
| OGIQUQKNJJTLSZ-UHFFFAOYSA-N | |
| 4-butylaniline | |
| 7694 | |
| ≥97.0% (GC,T) | |
| 4-Butylaniline |
| 250°C | |
| MFCD00007925 | |
| 4-n-butylaniline, benzenamine, 4-butyl, p-n-butylaniline, p-butylaniline, 4-butylbenzenamine, p-aminobutylbenzene, p-butylaminobenzene, 1-amino-4-butylbenzene, p-butyl aniline, 4-butylphenylamine | |
| CCCCC1=CC=C(C=C1)N | |
| 149.237 | |
| 149.24 | |
| Liquid |
Safety and Handling
EINECSNumber : (3)-0206
RTECSNumber : BW9470000
TSCA : Yes