accessibility menu, dialog, popup

UserName

H3CNH2

CAS RN 104-13-2

IUPAC Name: 4-butylaniline
Synonyms: benzenamine, 4-butyl-
Molecular Weight (g/mol): 149.24
Molecular Formula: C10H15N
InChi Key: OGIQUQKNJJTLSZ-UHFFFAOYSA-N
SMILES: CCCCC1=CC=C(N)C=C1
Boiling Point 
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)