Learn More
4-Butylaniline, 97%
CAS: 104-13-2 | C10H15N | 149.23 g/mol
$76.01 - $499.55
Chemical Identifiers
| CAS | 104-13-2 |
|---|---|
| Molecular Formula | C10H15N |
| Molecular Weight (g/mol) | 149.23 |
| InChI Key | OGIQUQKNJJTLSZ-UHFFFAOYSA-N |
| Synonym | 4-n-butylaniline, benzenamine, 4-butyl, p-n-butylaniline, p-butylaniline, 4-butylbenzenamine, p-aminobutylbenzene, p-butylaminobenzene, 1-amino-4-butylbenzene, p-butyl aniline, 4-butylphenylamine |
| PubChem CID | 7694 |
| IUPAC Name | 4-butylaniline |
| SMILES | CCCCC1=CC=C(C=C1)N |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC150650250
|
Thermo Scientific Chemicals
150650250 |
25 g | Glass bottle |
Each for $76.01
|
|
||||
|
AC150651000
|
Thermo Scientific Chemicals
150651000 |
100 g | Glass bottle |
Each for $170.08
|
|
||||
|
AC150655000
|
Thermo Scientific Chemicals
150655000 |
500 g | Glass bottle |
Each for $499.55
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 104-13-2 | |
| 149.23 | |
| 4-n-butylaniline, benzenamine, 4-butyl, p-n-butylaniline, p-butylaniline, 4-butylbenzenamine, p-aminobutylbenzene, p-butylaminobenzene, 1-amino-4-butylbenzene, p-butyl aniline, 4-butylphenylamine | |
| 4-butylaniline |
| C10H15N | |
| OGIQUQKNJJTLSZ-UHFFFAOYSA-N | |
| 7694 | |
| CCCCC1=CC=C(C=C1)N |
Specifications
| 104-13-2 | |
| 100.0 | |
| 0.9450g/mL | |
| 101°C | |
| 96% min. (GC) | |
| C10H15N | |
| CH3(CH2)3C6H4NH2 | |
| 12,I,503 | |
| 4-n-butylaniline, benzenamine, 4-butyl, p-n-butylaniline, p-butylaniline, 4-butylbenzenamine, p-aminobutylbenzene, p-butylaminobenzene, 1-amino-4-butylbenzene, p-butyl aniline, 4-butylphenylamine | |
| CCCCC1=CC=C(C=C1)N | |
| 149.23 | |
| 149.23 | |
| Liquid |
| 96.0 | |
| Brown to Yellow | |
| 133.0°C to 134.0°C (14.0 mmHg) | |
| Authentic | |
| Glass bottle | |
| 1.535 | |
| 25 g | |
| 0.945 | |
| OGIQUQKNJJTLSZ-UHFFFAOYSA-N | |
| 4-butylaniline | |
| 7694 | |
| 97% | |
| 4-Butylaniline |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
EINECSNumber : 203-177-4
RUO – Research Use Only