missing translation for 'onlineSavingsMsg'
Learn More

4-Butoxyphenylacetic Acid 98.0+%, TCI America™

Catalog No. p-7123716
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B47421G 1 g
B47425G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B47421G Supplier TCI America Supplier No. B47421G

Chemical Identifiers

CAS 4547-57-3
Molecular Formula C12H16O3
Molecular Weight (g/mol) 208.257
MDL Number MFCD00016832
InChI Key KLJMYYFCWBVKEE-UHFFFAOYSA-N
Synonym 4-butoxyphenylacetic acid, 4-n-butoxyphenylacetic acid, benzeneacetic acid, 4-butoxy, 2-4-butoxyphenyl acetic acid, acetic acid, p-butoxyphenyl, 4-butoxyphenyl acetic acid, 4-butoxybenzeneacetic acid, p-butoxyphenyl acetic acid, 4-butoxy-phenyl-acetic acid, p-butoxyphenylacetic acid
PubChem CID 78304
IUPAC Name 2-(4-butoxyphenyl)acetic acid
SMILES CCCCOC1=CC=C(C=C1)CC(=O)O

Specifications

Melting Point 87°C
Color White-Yellow
Quantity 1 g
Formula Weight 208.26
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Butoxyphenylacetic Acid
RTECSNumber AF6945000
TSCA No