missing translation for 'onlineSavingsMsg'
Learn More

4-Butoxyphenol 97.0+%, TCI America™

Catalog No. B0923500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0923500G 500 g
B092325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B0923500G Supplier TCI America Supplier No. B0923500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 122-94-1
Molecular Formula C10H14O2
Molecular Weight (g/mol) 166.22
MDL Number MFCD00002336
InChI Key MBGGFXOXUIDRJD-UHFFFAOYSA-N
Synonym phenol, 4-butoxy, p-butoxyphenol, 4-n-butoxyphenol, phenol, p-butoxy, hydroquinone monobutyl ether, unii-8188dbs68j, 4-normal-butoxyphenol, 4-butoxy-pheno, 4-butoxy-phenol, 4-n-butyloxyphenol
PubChem CID 31233
ChEBI CHEBI:34392
IUPAC Name 4-butoxyphenol
SMILES CCCCOC1=CC=C(O)C=C1

Specifications

Melting Point 66°C
Boiling Point 165°C
Quantity 500 g
Formula Weight 166.22
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Butoxyphenol
EINECSNumber (3)-3539
TSCA Yes