missing translation for 'onlineSavingsMsg'
Learn More

4-Butoxybenzaldehyde, 99%, Thermo Scientific™

$188.68 - $188.68

Chemical Identifiers

CAS 5736-88-9
Molecular Formula C11H14O2
Molecular Weight (g/mol) 178.23
MDL Number MFCD00003389
InChI Key XHWMNHADTZZHGI-UHFFFAOYSA-N
Synonym p-butoxybenzaldehyde, benzaldehyde, 4-butoxy, 4-n-butoxybenzaldehyde, benzaldehyde, p-butoxy, 4-butoxy-benzaldehyde, p-n-butoxybenzaldehyde, 4-n-butyloxybenzaldehyde, acmc-209lxn, p-n-butoxy benzaldehyde, 4-butoxybenzaldehyde
PubChem CID 79813
IUPAC Name 4-butoxybenzaldehyde
SMILES CCCCOC1=CC=C(C=C1)C=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC218370100
SDS View Documents
Thermo Scientific Chemicals
218370100
10 g Glass bottle
Each for $188.68
Only null left
 
Description

Description

Specifications

Chemical Identifiers

5736-88-9
178.23
XHWMNHADTZZHGI-UHFFFAOYSA-N
79813
CCCCOC1=CC=C(C=C1)C=O
C11H14O2
MFCD00003389
p-butoxybenzaldehyde, benzaldehyde, 4-butoxy, 4-n-butoxybenzaldehyde, benzaldehyde, p-butoxy, 4-butoxy-benzaldehyde, p-n-butoxybenzaldehyde, 4-n-butyloxybenzaldehyde, acmc-209lxn, p-n-butoxy benzaldehyde, 4-butoxybenzaldehyde
4-butoxybenzaldehyde

Specifications

5736-88-9
100.0
148°C to 149°C (10.0 mmHg)
99%
C11H14O2
CH3(CH2)3OC6H4CHO
10 g
p-butoxybenzaldehyde, benzaldehyde, 4-butoxy, 4-n-butoxybenzaldehyde, benzaldehyde, p-butoxy, 4-butoxy-benzaldehyde, p-n-butoxybenzaldehyde, 4-n-butyloxybenzaldehyde, acmc-209lxn, p-n-butoxy benzaldehyde, 4-butoxybenzaldehyde
CCCCOC1=CC=C(C=C1)C=O
178.23
178.23
Liquid
98.5
Red to Yellow
Authentic
Glass bottle
1.5390 to 1.541
MFCD00003389
08, II, 67
XHWMNHADTZZHGI-UHFFFAOYSA-N
4-butoxybenzaldehyde
79813
99%
4-Butoxybenzaldehyde
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 227-247-9

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only