accessibility menu, dialog, popup

UserName

H3COO

CAS RN 5736-88-9

IUPAC Name: 4-butoxybenzaldehyde
Synonyms: 4-butoxybenzaldehyde
Molecular Weight (g/mol): 178.23
Molecular Formula: C11H14O2
InChi Key: XHWMNHADTZZHGI-UHFFFAOYSA-N
SMILES: CCCCOC1=CC=C(C=O)C=C1
Boiling Point 
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)