missing translation for 'onlineSavingsMsg'
Learn More

4-Butoxybenzaldehyde, 99%, Thermo Scientific™

Catalog No. AC218370100
Change view
Click to view available options
Quantity:
10 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC218370100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC218370100 Supplier Thermo Scientific Chemicals Supplier No. 218370100
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5736-88-9
Molecular Formula C11H14O2
Molecular Weight (g/mol) 178.23
MDL Number MFCD00003389
InChI Key XHWMNHADTZZHGI-UHFFFAOYSA-N
Synonym p-butoxybenzaldehyde, benzaldehyde, 4-butoxy, 4-n-butoxybenzaldehyde, benzaldehyde, p-butoxy, 4-butoxy-benzaldehyde, p-n-butoxybenzaldehyde, 4-n-butyloxybenzaldehyde, acmc-209lxn, p-n-butoxy benzaldehyde, 4-butoxybenzaldehyde
PubChem CID 79813
IUPAC Name 4-butoxybenzaldehyde
SMILES CCCCOC1=CC=C(C=C1)C=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Red to Yellow
Boiling Point 148°C to 149°C (10.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.5390 to 1.541
Linear Formula CH3(CH2)3OC6H4CHO
Quantity 10 g
Beilstein 08, II, 67
Formula Weight 178.23
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 4-Butoxybenzaldehyde
Show More Show Less
EINECSNumber 227-247-9

RUO – Research Use Only