missing translation for 'onlineSavingsMsg'
Learn More

4-Butoxyaniline 98.0+%, TCI America™

Catalog No. B10035G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B10035G 5 g
B100325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B10035G Supplier TCI America Supplier No. B10035G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4344-55-2
Molecular Formula C10H15NO
Molecular Weight (g/mol) 165.236
MDL Number MFCD00007866
InChI Key UBRIHZOFEJHMIT-UHFFFAOYSA-N
Synonym p-butoxyaniline, benzenamine, 4-butoxy, p-butyloxyaniline, 4-n-butoxyaniline, aniline, p-butoxy, p-aminophenol n-butyl ether, 4-butyloxybenzenamine, aminophenyl-n-butyl ether, 4-butyloxyaniline, 4-butyloxy aniline
PubChem CID 20352
IUPAC Name 4-butoxyaniline
SMILES CCCCOC1=CC=C(C=C1)N

Specifications

Color Brown
Boiling Point 157°C
Quantity 5 g
Formula Weight 165.24
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-Butoxyaniline