Learn More
4-Bromotoluene, 98%
CAS: 106-38-7 | C7H7Br | 171.04 g/mol
Supplier: Thermo Scientific Chemicals A1584314
Description
4-Bromotoluene was used in the synthesis of ketones by Cu/Pd-catalyzed decarboxylative cross-coupling reaction. 4-Bromotoluene undergoes Heck reaction with styrene in the presence of in situ generated palladium complexes of phosphine-functionalized N-heterocyclic carbene ligands.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Bromotoluene was used in the synthesis of ketones by Cu/Pd-catalyzed decarboxylative cross-coupling reaction. 4-Bromotoluene undergoes Heck reaction with styrene in the presence of in situ generated palladium complexes of phosphine-functionalized N-heterocyclic carbene ligands.
Solubility
Insoluble in water. Soluble in methanol.
Notes
Store at room temperature. Keep away from oxidizing agents.
Specifications
| 4-Bromotoluene | |
| 26°C to 29°C | |
| 183°C to 184°C | |
| C7H7Br | |
| 25 g | |
| 14,1439 | |
| Insoluble in water. Soluble in methanol. | |
| CC1=CC=C(Br)C=C1 | |
| 171.04 | |
| 171.04 |
| 106-38-7 | |
| 1.41 | |
| 85°C (185°F) | |
| MFCD00000109 | |
| 1903636 | |
| 4-bromotoluene, p-bromotoluene, 4-methylbromobenzene, p-tolyl bromide, benzene, 1-bromo-4-methyl, toluene, p-bromo, p-methylphenyl bromide, p-methylbromobenzene, 1-methyl-4-bromobenzene, 4-bromo-1-methylbenzene | |
| ZBTMRBYMKUEVEU-UHFFFAOYSA-N | |
| 1-bromo-4-methylbenzene | |
| 7805 | |
| 98% |
Chemical Identifiers
| 106-38-7 | |
| 171.04 | |
| ZBTMRBYMKUEVEU-UHFFFAOYSA-N | |
| 7805 | |
| CC1=CC=C(Br)C=C1 |
| C7H7Br | |
| MFCD00000109 | |
| 4-bromotoluene, p-bromotoluene, 4-methylbromobenzene, p-tolyl bromide, benzene, 1-bromo-4-methyl, toluene, p-bromo, p-methylphenyl bromide, p-methylbromobenzene, 1-methyl-4-bromobenzene, 4-bromo-1-methylbenzene | |
| 1-bromo-4-methylbenzene |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 203-391-8
RTECSNumber : XS7965600
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only