missing translation for 'onlineSavingsMsg'
Learn More

4-Bromothiophene-2-carboxaldehyde 98.0+%, TCI America™

Catalog No. p-7119786
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
B28275G 5 g
B282725G 25 g
2 options

Catalog No. B28275G

Supplier: TCI America B28275G

Only null left

Chemical Identifiers

CAS 18791-75-8
Molecular Formula C5H3BrOS
Molecular Weight (g/mol) 191.04
MDL Number MFCD00005431
InChI Key PDONIKHDXYHTLS-UHFFFAOYSA-N
Synonym 4-bromothiophene-2-carboxaldehyde, 4-bromo-2-thiophenecarboxaldehyde, 4-bromo-2-formylthiophene, 4-bromo-2-thiophenecarbaldehyde, 4-bromo-thiophene-2-carbaldehyde, 4-bromo-2-thiophene carboxaldehyde, 4-bromothiophen-2-carboxaldehyde, 2-thiophenecarboxaldehyde, 4-bromo, 4-bromothiophene-2-aldehyde, 3-bromothiophene-5-carboxaldehyde
PubChem CID 87792
IUPAC Name 4-bromothiophene-2-carbaldehyde
SMILES BrC1=CSC(C=O)=C1

Specifications

Melting Point 46°C
Boiling Point 115°C
Quantity 5 g
Formula Weight 191.04
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromothiophene-2-carboxaldehyde