missing translation for 'onlineSavingsMsg'
Learn More

4-Bromothiophene-2-carboxaldehyde, 96%

Catalog No. AAA1330406
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1330406 5 g
AAA1330414 25 g
AAA1330422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1330406 Supplier Thermo Scientific Chemicals Supplier No. A1330406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 18791-75-8 | C5H3BrOS | 191.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 18791-75-8
Molecular Formula C5H3BrOS
Molecular Weight (g/mol) 191.04
MDL Number MFCD00005431
InChI Key PDONIKHDXYHTLS-UHFFFAOYSA-N
Synonym 4-bromothiophene-2-carboxaldehyde, 4-bromo-2-thiophenecarboxaldehyde, 4-bromo-2-formylthiophene, 4-bromo-2-thiophenecarbaldehyde, 4-bromo-thiophene-2-carbaldehyde, 4-bromo-2-thiophene carboxaldehyde, 4-bromothiophen-2-carboxaldehyde, 2-thiophenecarboxaldehyde, 4-bromo, 4-bromothiophene-2-aldehyde, 3-bromothiophene-5-carboxaldehyde
PubChem CID 87792
IUPAC Name 4-bromothiophene-2-carbaldehyde
SMILES BrC1=CSC(C=O)=C1

Specifications

Melting Point 44°C to 48°C
Boiling Point 114°C to 115°C (11 mmHg)
Flash Point >110°C (230°F)
Quantity 5 g
Sensitivity Air sensitive
Formula Weight 191.05
Percent Purity 96%
Chemical Name or Material 4-Bromothiophene-2-carboxaldehyde
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 242-577-3
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only