missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromothioanisole 99.0+%, TCI America™
$200.21 - $200.21
Chemical Identifiers
| CAS | 104-95-0 |
|---|---|
| Molecular Formula | C7H7BrS |
| Molecular Weight (g/mol) | 203.10 |
| MDL Number | MFCD00000102 |
| InChI Key | YEUYZNNBXLMFCW-UHFFFAOYSA-N |
| Synonym | 4-bromothioanisole, 1-bromo-4-methylthio benzene, p-bromophenyl methyl sulfide, 4-bromophenyl methyl sulfide, benzene, 1-bromo-4-methylthio, 1-bromo-4-methylsulfanyl benzene, p-bromothioanisole, 4-bromo-thioanisole, methyl 4-bromophenylsulfide, p-bromo thioanisole |
| PubChem CID | 66037 |
| IUPAC Name | 1-bromo-4-(methylsulfanyl)benzene |
| SMILES | CSC1=CC=C(Br)C=C1 |
Chemical Identifiers
| 104-95-0 | |
| 203.10 | |
| YEUYZNNBXLMFCW-UHFFFAOYSA-N | |
| 66037 | |
| CSC1=CC=C(Br)C=C1 |
| C7H7BrS | |
| MFCD00000102 | |
| 4-bromothioanisole, 1-bromo-4-methylthio benzene, p-bromophenyl methyl sulfide, 4-bromophenyl methyl sulfide, benzene, 1-bromo-4-methylthio, 1-bromo-4-methylsulfanyl benzene, p-bromothioanisole, 4-bromo-thioanisole, methyl 4-bromophenylsulfide, p-bromo thioanisole | |
| 1-bromo-4-(methylsulfanyl)benzene |
Specifications
| 104-95-0 | |
| White-Yellow | |
| C7H7BrS | |
| 25 g | |
| YEUYZNNBXLMFCW-UHFFFAOYSA-N | |
| 1-bromo-4-(methylsulfanyl)benzene | |
| 66037 | |
| ≥99.0% (GC) | |
| 4-Bromothioanisole |
| 38°C | |
| 250°C | |
| MFCD00000102 | |
| 4-bromothioanisole, 1-bromo-4-methylthio benzene, p-bromophenyl methyl sulfide, 4-bromophenyl methyl sulfide, benzene, 1-bromo-4-methylthio, 1-bromo-4-methylsulfanyl benzene, p-bromothioanisole, 4-bromo-thioanisole, methyl 4-bromophenylsulfide, p-bromo thioanisole | |
| CSC1=CC=C(Br)C=C1 | |
| 203.10 | |
| 203.10 | |
| Crystalline Powder |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator