missing translation for 'onlineSavingsMsg'
Learn More

4-Bromothioanisole 99.0+%, TCI America™

Catalog No. B202425G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B202425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B202425G Supplier TCI America Supplier No. B202425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 104-95-0
Molecular Formula C7H7BrS
Molecular Weight (g/mol) 203.10
MDL Number MFCD00000102
InChI Key YEUYZNNBXLMFCW-UHFFFAOYSA-N
Synonym 4-bromothioanisole, 1-bromo-4-methylthio benzene, p-bromophenyl methyl sulfide, 4-bromophenyl methyl sulfide, benzene, 1-bromo-4-methylthio, 1-bromo-4-methylsulfanyl benzene, p-bromothioanisole, 4-bromo-thioanisole, methyl 4-bromophenylsulfide, p-bromo thioanisole
PubChem CID 66037
IUPAC Name 1-bromo-4-(methylsulfanyl)benzene
SMILES CSC1=CC=C(Br)C=C1

Specifications

Melting Point 38°C
Color White-Yellow
Boiling Point 250°C
Quantity 25 g
Formula Weight 203.10
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromothioanisole
TSCA No
Recommended Storage Refrigerator