missing translation for 'onlineSavingsMsg'
Learn More

4-Bromothiazole 98.0+%, TCI America™

Catalog No. p-7123633
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B45941G 1 g
B45945G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B45941G Supplier TCI America Supplier No. B45941G
Only null left

Chemical Identifiers

CAS 34259-99-9
Molecular Formula C3H2BrNS
Molecular Weight (g/mol) 164.02
MDL Number MFCD06657592
InChI Key VDTIGYKLTROQAH-UHFFFAOYSA-N
Synonym 4-bromothiazole, 4-bromo-thiazole, 4-bromo thiazole, thiazole, 4-bromo, 4-bromothiazol, zlchem 498, pubchem4040, ksc222g1h
PubChem CID 2763218
IUPAC Name 4-bromo-1,3-thiazole
SMILES BrC1=CSC=N1

Specifications

Color Yellow
Boiling Point 190°C
Quantity 1 g
Formula Weight 164.02
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Bromothiazole