accessibility menu, dialog, popup

UserName

CAS RN 34259-99-9

IUPAC Name: 4-bromo-1,3-thiazole
Synonyms: 4-bromo-1,3-thiazole
Molecular Weight (g/mol): 164.02
Molecular Formula: C3H2BrNS
InChi Key: VDTIGYKLTROQAH-UHFFFAOYSA-N
SMILES: BrC1=CSC=N1
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)