missing translation for 'onlineSavingsMsg'
Learn More

4-Bromothiazole, 97%, Thermo Scientific™

Chemical Identifiers

CAS 34259-99-9
Molecular Formula C3H2BrNS
Molecular Weight (g/mol) 164.02
MDL Number MFCD06657592
InChI Key VDTIGYKLTROQAH-UHFFFAOYSA-N
Synonym 4-bromothiazole, 4-bromo-thiazole, 4-bromo thiazole, thiazole, 4-bromo, 4-bromothiazol, zlchem 498, pubchem4040, ksc222g1h
PubChem CID 2763218
IUPAC Name 4-bromo-1,3-thiazole
SMILES BrC1=CSC=N1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC429450010
SDS View Documents
Thermo Scientific Chemicals
429450010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC429450050
SDS View Documents
Thermo Scientific Chemicals
429450050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

34259-99-9
164.02
VDTIGYKLTROQAH-UHFFFAOYSA-N
2763218
BrC1=CSC=N1
C3H2BrNS
MFCD06657592
4-bromothiazole, 4-bromo-thiazole, 4-bromo thiazole, thiazole, 4-bromo, 4-bromothiazol, zlchem 498, pubchem4040, ksc222g1h
4-bromo-1,3-thiazole

Specifications

34259-99-9
100.0
Authentic
Glass bottle
MFCD06657592
4-bromothiazole, 4-bromo-thiazole, 4-bromo thiazole, thiazole, 4-bromo, 4-bromothiazol, zlchem 498, pubchem4040, ksc222g1h
BrC1=CSC=N1
164.02
164.02
4-Bromothiazole
96.0
102°C
96% min. (GC)
C3H2BrNS
1g
VDTIGYKLTROQAH-UHFFFAOYSA-N
4-bromo-1,3-thiazole
2763218
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/e

GHS Signal Word: Warning

SDS
Documents

Documents

RUO – Research Use Only