missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromotetrahydro-2H-pyran 98.0+%, TCI America™
$126.76 - $372.81
Chemical Identifiers
| CAS | 25637-16-5 |
|---|---|
| Molecular Formula | C5H9BrO |
| Molecular Weight (g/mol) | 165.03 |
| MDL Number | MFCD07371501 |
| InChI Key | IVBVKTPDEWDNRW-UHFFFAOYSA-N |
| PubChem CID | 13349654 |
| IUPAC Name | 4-bromooxane |
| SMILES | C1COCCC1Br |
Chemical Identifiers
| 25637-16-5 | |
| 165.03 | |
| IVBVKTPDEWDNRW-UHFFFAOYSA-N | |
| 4-bromooxane |
| C5H9BrO | |
| MFCD07371501 | |
| 13349654 | |
| C1COCCC1Br |
Specifications
| 25637-16-5 | |
| C5H9BrO | |
| 1 g | |
| C1COCCC1Br | |
| 165.03 | |
| 165.03 | |
| Liquid |
| 61°C | |
| MFCD07371501 | |
| IVBVKTPDEWDNRW-UHFFFAOYSA-N | |
| 4-bromooxane | |
| 13349654 | |
| ≥98.0% (GC) | |
| 4-Bromotetrahydro-2H-pyran |
Safety and Handling
TSCA : No