Learn More
4-Bromostyrene, 98%, stab. with 0.1% 4-tert-butylcatechol
CAS: 2039-82-9 | C8H7Br | 183.05 g/mol
Supplier: Thermo Scientific Chemicals A1789606
Description
4-Bromostyrene is widely utilized in the synthesis of nitroolefins by alkene cross-metathesis. It plays an important role in the Heck reaction to the synthesis of poly(1,4-phenylenevinylene). It is employed in the structure activity relationships (SAR) study of the chemical and biochemical properties of the vinyl group of styrene. It is also used to investigate the photochemical growth of Br-terminated self-assembled monolayers. It is also involved in the synthesis of silsesquioxanes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Bromostyrene is widely utilized in the synthesis of nitroolefins by alkene cross-metathesis. It plays an important role in the Heck reaction to the synthesis of poly(1,4-phenylenevinylene). It is employed in the structure activity relationships (SAR) study of the chemical and biochemical properties of the vinyl group of styrene. It is also used to investigate the photochemical growth of Br-terminated self-assembled monolayers. It is also involved in the synthesis of silsesquioxanes.
Solubility
Miscible with ethanol, ether and benzene.
Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with acids, bases, oxidizing agents and halogens. Keep away from sources of ignition.
Specifications
| 4-Bromostyrene | |
| 2039-82-9 | |
| 1.4 | |
| 75°C (167°F) | |
| 1.594 | |
| 5 g | |
| 4-bromostyrene, 1-bromo-4-vinylbenzene, p-bromostyrene, benzene, 1-bromo-4-ethenyl, styrene, p-bromo, 1-4-bromophenyl ethylene, unii-tvk4s80xlu, p-bromo styrene, 1-bromo-4-vinyl-benzene, tvk4s80xlu | |
| WGGLDBIZIQMEGH-UHFFFAOYSA-N | |
| 1-bromo-4-ethenylbenzene | |
| 16263 | |
| 98% |
| stabilized with 0.1% 4-tert-butylcatechol | |
| 3°C to 6°C | |
| 88°C to 89°C (16 mmHg) | |
| C8H7Br | |
| MFCD00000110 | |
| 1634204 | |
| Miscible with ethanol,ether and benzene. | |
| BrC1=CC=C(C=C)C=C1 | |
| 183.05 | |
| 183.05 |
Chemical Identifiers
| 2039-82-9 | |
| 183.05 | |
| WGGLDBIZIQMEGH-UHFFFAOYSA-N | |
| 16263 | |
| BrC1=CC=C(C=C)C=C1 |
| C8H7Br | |
| MFCD00000110 | |
| 4-bromostyrene, 1-bromo-4-vinylbenzene, p-bromostyrene, benzene, 1-bromo-4-ethenyl, styrene, p-bromo, 1-4-bromophenyl ethylene, unii-tvk4s80xlu, p-bromo styrene, 1-bromo-4-vinyl-benzene, tvk4s80xlu | |
| 1-bromo-4-ethenylbenzene |
Safety and Handling
GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P302+P352-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P374-P501c
H227-H315-H319-H335
EINECSNumber : 218-022-6
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only