missing translation for 'onlineSavingsMsg'
Learn More

4-Bromostyrene, 96%, stabilized

Catalog No. AC154280100
Change view
Click to view available options
Quantity:
10 mL
25 mL
100 mL
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC154280100 10 mL Glass bottle
AC154280250 25 mL Glass bottle
AC154281000 100 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC154280100 Supplier Thermo Scientific Chemicals Supplier No. 154280100

CAS: 2039-82-9 | C8H7Br | 183.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2039-82-9
Molecular Formula C8H7Br
Molecular Weight (g/mol) 183.05
MDL Number MFCD00000110
InChI Key WGGLDBIZIQMEGH-UHFFFAOYSA-N
Synonym 4-bromostyrene, 1-bromo-4-vinylbenzene, p-bromostyrene, benzene, 1-bromo-4-ethenyl, styrene, p-bromo, 1-4-bromophenyl ethylene, unii-tvk4s80xlu, p-bromo styrene, 1-bromo-4-vinyl-benzene, tvk4s80xlu
PubChem CID 16263
IUPAC Name 1-bromo-4-ethenylbenzene
SMILES BrC1=CC=C(C=C)C=C1

Specifications

Melting Point 4.5°C
Color Colorless to Yellow
Density 1.4000g/mL
Boiling Point 88.0°C to 89.0°C (16.0 mmHg)
Flash Point 75°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Refractive Index 1.5930 to 1.5950
Linear Formula H2C=CHC6H4Br
Quantity 10 mL
Beilstein 05,II,367
Specific Gravity 1.4
Stabilizer 100 to 500ppm 4-tert.-butylcatechol
Formula Weight 183.05
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 4-Bromostyrene
Show More Show Less
Hazard Category GHS Signal Word: Warning
EINECSNumber 218-022-6

RUO – Research Use Only