missing translation for 'onlineSavingsMsg'
Learn More

4-Bromosalicylaldehyde 98.0+%, TCI America™

Catalog No. p-7123064
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B36371G 1 g
B36375G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B36371G Supplier TCI America Supplier No. B36371G
Only null left

Chemical Identifiers

CAS 22532-62-3
Molecular Formula C7H5BrO2
Molecular Weight (g/mol) 201.019
MDL Number MFCD06252606
InChI Key HXTWKHXDFATMSP-UHFFFAOYSA-N
Synonym 4-bromosalicylaldehyde, 4-bromo-2-hydroxy-benzaldehyde, 2-hydroxy-4-bromobenzaldehyde, benzaldehyde, 4-bromo-2-hydroxy, 4-bromo-2-hydroxybenzaldheyde, 4-bromosalicyladehyde, pubchem16944, acmc-1cq5t, intermediates-zcf02605, 4-bromo-2-hydroxybenaldehyde
PubChem CID 4066019
IUPAC Name 4-bromo-2-hydroxybenzaldehyde
SMILES C1=CC(=C(C=C1Br)O)C=O

Specifications

Melting Point 53°C
Quantity 1 g
Formula Weight 201.02
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromosalicylaldehyde