missing translation for 'onlineSavingsMsg'
Learn More

4-Bromosalicylaldehyde, 97%

Catalog No. p-7036541
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5485003 1 g
AAH5485006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5485003 Supplier Thermo Scientific Chemicals Supplier No. H5485003

CAS: 22532-62-3 | C7H5BrO2 | 201.019 g/mol

It is an important raw material and intermediate used in organic synthesis, Pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, Pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Soluble in methanol.

Notes
Strong bases and oxidizing agents are to be avoided.

Chemical Identifiers

CAS 22532-62-3
Molecular Formula C7H5BrO2
Molecular Weight (g/mol) 201.019
MDL Number MFCD06252606
InChI Key HXTWKHXDFATMSP-UHFFFAOYSA-N
Synonym 4-bromosalicylaldehyde, 4-bromo-2-hydroxy-benzaldehyde, 2-hydroxy-4-bromobenzaldehyde, benzaldehyde, 4-bromo-2-hydroxy, 4-bromo-2-hydroxybenzaldheyde, 4-bromosalicyladehyde, pubchem16944, acmc-1cq5t, intermediates-zcf02605, 4-bromo-2-hydroxybenaldehyde
PubChem CID 4066019
IUPAC Name 4-bromo-2-hydroxybenzaldehyde
SMILES C1=CC(=C(C=C1Br)O)C=O

Specifications

Melting Point 50°C to 54°C
Flash Point >110°C (230°F)
Quantity 1 g
UN Number UN3077
Solubility Information Soluble in methanol.
Formula Weight 201.02
Percent Purity 97%
Chemical Name or Material 4-Bromosalicylaldehyde
Hazard Category H302-H319
Hazard Statement GHS H Statement
H301-H318
Toxic if swallowed.
Causes serious eye damage.
Precautionary Statement P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only