missing translation for 'onlineSavingsMsg'
Learn More

4-Bromoresorcinol 98.0+%, TCI America™

Catalog No. B06545G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B06545G 5 g
B065425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B06545G Supplier TCI America Supplier No. B06545G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6626-15-9
Molecular Formula C6H5BrO2
Molecular Weight (g/mol) 189.008
MDL Number MFCD00002272
InChI Key MPCCNXGZCOXPMG-UHFFFAOYSA-N
Synonym 4-bromoresorcinol, 1-bromo-2,4-dihydroxybenzene, 1,3-benzenediol, 4-bromo, 4-bromo-1,3-dihydroxybenzene, 4-bromo-benzene-1,3-diol, 4-bromo-resorcinol, resorcinol, 4-bromo, acmc-209nuf, 2,4-dihydroxybromobenzene
PubChem CID 81105
IUPAC Name 4-bromobenzene-1,3-diol
SMILES C1=CC(=C(C=C1O)O)Br

Specifications

Melting Point 103°C
Boiling Point 150°C
Quantity 5 g
Formula Weight 189.01
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromoresorcinol