missing translation for 'onlineSavingsMsg'
Learn More

4-Bromoresorcinol, 98%

Catalog No. AC107430050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC107430050 5 g Glass bottle
AC107430250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC107430050 Supplier Thermo Scientific Chemicals Supplier No. 107430050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 6626-15-9 | C6H5BrO2 | 189.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6626-15-9
Molecular Formula C6H5BrO2
Molecular Weight (g/mol) 189.01
MDL Number MFCD00002272
InChI Key MPCCNXGZCOXPMG-UHFFFAOYSA-N
Synonym 4-bromoresorcinol, 1-bromo-2,4-dihydroxybenzene, 1,3-benzenediol, 4-bromo, 4-bromo-1,3-dihydroxybenzene, 4-bromo-benzene-1,3-diol, 4-bromo-resorcinol, resorcinol, 4-bromo, acmc-209nuf, 2,4-dihydroxybromobenzene
PubChem CID 81105
IUPAC Name 4-bromobenzene-1,3-diol
SMILES C1=CC(=C(C=C1O)O)Br

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 99°C to 102°C
Color Beige to Pink
Boiling Point 150°C (12.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula BrC6H3(OH)2
Quantity 5 g
Beilstein 06, 821
Formula Weight 189.01
Percent Purity 98%
Physical Form Crystals or Powder
Chemical Name or Material 4-Bromoresorcinol
Show More Show Less
EINECSNumber 229-586-8

RUO – Research Use Only