missing translation for 'onlineSavingsMsg'
Learn More

4-Bromopyrene 95.0+%, TCI America™

Catalog No. B28071G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B28071G 1 g
B28075G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B28071G Supplier TCI America Supplier No. B28071G
Only null left

Chemical Identifiers

CAS 1732-26-9
Molecular Formula C16H9Br
Molecular Weight (g/mol) 281.152
InChI Key QMHTZTOPYZKQLC-UHFFFAOYSA-N
PubChem CID 3014036
IUPAC Name 4-bromopyrene
SMILES C1=CC2=C3C(=C1)C=C(C4=CC=CC(=C43)C=C2)Br

Specifications

Melting Point 152°C
Color Yellow
Quantity 1 g
Formula Weight 281.15
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromopyrene