missing translation for 'onlineSavingsMsg'
Learn More

4'-Bromopropiophenone 99.0+%, TCI America™

Catalog No. B188725G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B188725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B188725G Supplier TCI America Supplier No. B188725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10342-83-3
Molecular Formula C9H9BrO
Molecular Weight (g/mol) 213.07
MDL Number MFCD00000106
InChI Key UOMOSYFPKGQIKI-UHFFFAOYSA-N
Synonym 4'-bromopropiophenone, 1-4-bromophenyl propan-1-one, p-bromopropiophenone, 4-bromopropiophenone, propiophenone, 4'-bromo, 4-bromophenyl ethyl ketone, 1-propanone, 1-4-bromophenyl, 1-4-bromophenyl-1-propanone, aurora 22532
PubChem CID 66312
IUPAC Name 1-(4-bromophenyl)propan-1-one
SMILES CCC(=O)C1=CC=C(Br)C=C1

Specifications

Melting Point 46°C
Color White-Yellow
Boiling Point 138°C
Quantity 25 g
Formula Weight 213.07
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Bromopropiophenone