accessibility menu, dialog, popup

UserName

CAS RN 10342-83-3

CH3OBr

CAS RN 10342-83-3

IUPAC Name: 1-(4-bromophenyl)propan-1-one
Synonyms: 1-(4-bromophenyl)propan-1-one
Molecular Weight (g/mol): 213.07
Molecular Formula: C9H9BrO
InChi Key: UOMOSYFPKGQIKI-UHFFFAOYSA-N
SMILES: CCC(=O)C1=CC=C(Br)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results