missing translation for 'onlineSavingsMsg'
Learn More

4-Bromophthalimide 98.0+%, TCI America™

Catalog No. p-7119418
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B20095G 5 g
B200925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B20095G Supplier TCI America Supplier No. B20095G
Only null left

Chemical Identifiers

CAS 6941-75-9
Molecular Formula C8H4BrNO2
Molecular Weight (g/mol) 226.029
MDL Number MFCD00466049
InChI Key GNYICZVGHULCHE-UHFFFAOYSA-N
Synonym 4-bromophthalimide, 5-bromoisoindoline-1,3-dione, 5-bromo-isoindole-1,3-dione, 5-bromo-2,3-dihydro-1h-isoindole-1,3-dione, 4-bromo phthalimide, 5-bromo-1h-isoindole-1,3 2h-dione, 1h-isoindole-1,3 2h-dione, 5-bromo, 5-bromo-2h-isoindole-1,3-dione, 5-bromophthalimide, acmc-209o8b
PubChem CID 236018
IUPAC Name 5-bromoisoindole-1,3-dione
SMILES C1=CC2=C(C=C1Br)C(=O)NC2=O

Specifications

Melting Point 235°C
Color White-Yellow
Quantity 5 g
Formula Weight 226.03
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromophthalimide