accessibility menu, dialog, popup

UserName

BrOHNO

CAS RN 6941-75-9

IUPAC Name: 5-bromo-2,3-dihydro-1H-isoindole-1,3-dione
Synonyms: 5-bromo-2H-isoindole-1,3-dione
Molecular Weight (g/mol): 226.03
Molecular Formula: C8H4BrNO2
InChi Key: GNYICZVGHULCHE-UHFFFAOYSA-N
SMILES: BrC1=CC=C2C(=O)NC(=O)C2=C1
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)