missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Bromophthalic Anhydride, TCI America™

$82.50 - $780.70

Chemical Identifiers

CAS 86-90-8
Molecular Formula C8H3BrO3
Molecular Weight (g/mol) 227.013
MDL Number MFCD00191323
InChI Key BCKVHOUUJMYIAN-UHFFFAOYSA-N
PubChem CID 66590
IUPAC Name 5-bromo-2-benzofuran-1,3-dione
SMILES C1=CC2=C(C=C1Br)C(=O)OC2=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B169325G View Documents TCI America
B169325G
25g Each for $82.50
 
B1693500G View Documents TCI America
B1693500G
500g Each for $780.70
 
Specifications

Chemical Identifiers

86-90-8
227.013
BCKVHOUUJMYIAN-UHFFFAOYSA-N
5-bromo-2-benzofuran-1,3-dione
C8H3BrO3
MFCD00191323
66590
C1=CC2=C(C=C1Br)C(=O)OC2=O

Specifications

301°C
107°C
86-90-8
MFCD00191323
C1=CC2=C(C=C1Br)C(=O)OC2=O
227.013
227.01
≥97.0% (GC)
White-Yellow
25g
C8H3BrO3
BCKVHOUUJMYIAN-UHFFFAOYSA-N
5-bromo-2-benzofuran-1,3-dione
66590
Crystal-Powder at 20°C
4-Bromophthalic Anhydride
Safety and Handling

Safety and Handling

TSCA : No