missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromophthalic Anhydride 97.0+%, TCI America™
Specifications
Specifications
| Melting Point | 107°C |
| Color | White-Yellow |
| Boiling Point | 301°C |
| Quantity | 25 g |
| Formula Weight | 227.01 |
| Percent Purity | ≥97.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4-Bromophthalic Anhydride |
Safety and Handling
| TSCA | No |