missing translation for 'onlineSavingsMsg'
Learn More

4-Bromophthalic Acid 98.0+%, TCI America™

Catalog No. B22571G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B22571G 1 g
B22575G 5 g
B225725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B22571G Supplier TCI America Supplier No. B22571G

Chemical Identifiers

CAS 6968-28-1
Molecular Formula C8H5BrO4
Molecular Weight (g/mol) 245.028
MDL Number MFCD00043538
InChI Key AZXKGUVDIORSED-UHFFFAOYSA-N
Synonym 4-bromophthalicacid, 4-bromobenzene-1,2-dicarboxylic acid, 1,2-benzenedicarboxylic acid, 4-bromo, zlchem 539, 5-bromo-phthalic acid, acmc-209oah, #, 4-bromo-1,2-benzenedicarboxylic acid, 1,2-benzenedicarboxylicacid, 4-bromo, 4-bromobenzene-1,2-dioic acid
PubChem CID 81428
IUPAC Name 4-bromophthalic acid
SMILES C1=CC(=C(C=C1Br)C(=O)O)C(=O)O

Specifications

Melting Point 166°C
Quantity 1 g
Formula Weight 245.03
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromophthalic Acid
EINECSNumber (3)-0062
TSCA No