missing translation for 'onlineSavingsMsg'
Learn More

4-Bromophthalic acid, 98%, Thermo Scientific Chemicals

$150.22 - $150.22

Chemical Identifiers

CAS 6968-28-1
Molecular Formula C8H5BrO4
Molecular Weight (g/mol) 245.028
MDL Number MFCD00043538
InChI Key AZXKGUVDIORSED-UHFFFAOYSA-N
Synonym 4-bromophthalicacid, 4-bromobenzene-1,2-dicarboxylic acid, 1,2-benzenedicarboxylic acid, 4-bromo, zlchem 539, 5-bromo-phthalic acid, acmc-209oah, #, 4-bromo-1,2-benzenedicarboxylic acid, 1,2-benzenedicarboxylicacid, 4-bromo, 4-bromobenzene-1,2-dioic acid
PubChem CID 81428
IUPAC Name 4-bromophthalic acid
SMILES C1=CC(=C(C=C1Br)C(=O)O)C(=O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1341806
View Documents
Thermo Scientific Chemicals
L1341806
5 g
Each for $150.22
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

6968-28-1
245.028
AZXKGUVDIORSED-UHFFFAOYSA-N
81428
C1=CC(=C(C=C1Br)C(=O)O)C(=O)O
C8H5BrO4
MFCD00043538
4-bromophthalicacid, 4-bromobenzene-1,2-dicarboxylic acid, 1,2-benzenedicarboxylic acid, 4-bromo, zlchem 539, 5-bromo-phthalic acid, acmc-209oah, #, 4-bromo-1,2-benzenedicarboxylic acid, 1,2-benzenedicarboxylicacid, 4-bromo, 4-bromobenzene-1,2-dioic acid
4-bromophthalic acid

Specifications

6968-28-1
C8H5BrO4
5 g
4-bromophthalicacid, 4-bromobenzene-1,2-dicarboxylic acid, 1,2-benzenedicarboxylic acid, 4-bromo, zlchem 539, 5-bromo-phthalic acid, acmc-209oah, #, 4-bromo-1,2-benzenedicarboxylic acid, 1,2-benzenedicarboxylicacid, 4-bromo, 4-bromobenzene-1,2-dioic acid
C1=CC(=C(C=C1Br)C(=O)O)C(=O)O
245.028
245.03
4-Bromophthalic acid
162°C to 164°C
MFCD00043538
1959086
AZXKGUVDIORSED-UHFFFAOYSA-N
4-bromophthalic acid
81428
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 230-183-4

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only