missing translation for 'onlineSavingsMsg'
Learn More

4-Bromophthalic acid, 98%, Thermo Scientific Chemicals

Catalog No. AAL1341806
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1341806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1341806 Supplier Thermo Scientific Chemicals Supplier No. L1341806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6968-28-1 | Molecular Formula: C8H5BrO4 | Molecular Weight: 245.028 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6968-28-1
Molecular Formula C8H5BrO4
Molecular Weight (g/mol) 245.028
MDL Number MFCD00043538
InChI Key AZXKGUVDIORSED-UHFFFAOYSA-N
Synonym 4-bromophthalicacid, 4-bromobenzene-1,2-dicarboxylic acid, 1,2-benzenedicarboxylic acid, 4-bromo, zlchem 539, 5-bromo-phthalic acid, acmc-209oah, #, 4-bromo-1,2-benzenedicarboxylic acid, 1,2-benzenedicarboxylicacid, 4-bromo, 4-bromobenzene-1,2-dioic acid
PubChem CID 81428
IUPAC Name 4-bromophthalic acid
SMILES C1=CC(=C(C=C1Br)C(=O)O)C(=O)O

Specifications

Melting Point 162°C to 164°C
Quantity 5 g
Beilstein 1959086
Formula Weight 245.03
Percent Purity 98%
Chemical Name or Material 4-Bromophthalic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 230-183-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only