missing translation for 'onlineSavingsMsg'
Learn More

4-Bromophenylurea, 97%

Catalog No. AAL1107406
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1107406 5 g
AAL1107403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1107406 Supplier Thermo Scientific Chemicals Supplier No. L1107406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1967-25-5 | C7H7BrN2O | 215.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1967-25-5
Molecular Formula C7H7BrN2O
Molecular Weight (g/mol) 215.05
MDL Number MFCD00025428
InChI Key PFQUUCXMPUNRLA-UHFFFAOYSA-N
Synonym 4-bromophenyl urea, 1-4-bromophenyl urea, n-4-bromophenyl urea, p-bromophenylurea, unii-n4k2d5wzsi, n4k2d5wzsi, urea, 4-bromophenyl, 4-bromophenyl-urea, acmc-209f0h, amino-n-4-bromophenyl amide
PubChem CID 16074
IUPAC Name (4-bromophenyl)urea
SMILES C1=CC(=CC=C1NC(=O)N)Br

Specifications

Quantity 5 g
Beilstein 2090140
Formula Weight 215.06
Percent Purity 97%
Chemical Name or Material 4-Bromophenylurea
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only