missing translation for 'onlineSavingsMsg'
Learn More

4-Bromophenylhydrazine Hydrochloride 98.0+%, TCI America™

Catalog No. B06355G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B06355G 5 g
B063525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B06355G Supplier TCI America Supplier No. B06355G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 622-88-8
Molecular Formula C6H8BrClN2
Molecular Weight (g/mol) 223.498
MDL Number MFCD00012941
InChI Key RGGOWBBBHWTTRE-UHFFFAOYSA-N
Synonym 4-bromophenylhydrazine hydrochloride, 4-bromophenyl hydrazine hydrochloride, 4-bromophenylhydrazine hcl, 4-bromo phenyl hydrazine hydrochloride, p-bromophenylhydrazine monohydrochloride, hydrazine, 4-bromophenyl-, monohydrochloride, 1-4-bromophenyl hydrazine hydrochloride, 4-bromophenylhydrazinehydrochloride, p-bromophenylhydrazine hydrochloride
PubChem CID 12157
IUPAC Name (4-bromophenyl)hydrazine;hydrochloride
SMILES C1=CC(=CC=C1NN)Br.Cl

Specifications

Melting Point 230°C
Quantity 5 g
Formula Weight 223.50
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromophenylhydrazine Hydrochloride
RTECSNumber MV0800000
TSCA Yes