missing translation for 'onlineSavingsMsg'
Learn More

4-Bromophenylhydrazine hydrochloride, 97%

Catalog No. AAA1559006
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1559006 5 g
AAA1559014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1559006 Supplier Thermo Scientific Chemicals Supplier No. A1559006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 622-88-8 | C6H8BrClN2 | 223.498 g/mol

4-Bromophenylhydrazine hydrochloride is used as a reagent in the preparation of acylsulfonamides, acylsulfamides and 9-bromo-2-ethenyl-7,12-dihydroindolo[3,2-d][1]benzazepin-6(5H)-one.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromophenylhydrazine hydrochloride is used as a reagent in the preparation of acylsulfonamides, acylsulfamides and 9-bromo-2-ethenyl-7,12-dihydroindolo[3,2-d][1]benzazepin-6(5H)-one.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents and bases.

Chemical Identifiers

CAS 622-88-8
Molecular Formula C6H8BrClN2
Molecular Weight (g/mol) 223.498
MDL Number MFCD00012941
InChI Key RGGOWBBBHWTTRE-UHFFFAOYSA-N
Synonym 4-bromophenylhydrazine hydrochloride, 4-bromophenyl hydrazine hydrochloride, 4-bromophenylhydrazine hcl, 4-bromo phenyl hydrazine hydrochloride, p-bromophenylhydrazine monohydrochloride, hydrazine, 4-bromophenyl-, monohydrochloride, 1-4-bromophenyl hydrazine hydrochloride, 4-bromophenylhydrazinehydrochloride, p-bromophenylhydrazine hydrochloride
PubChem CID 12157
IUPAC Name (4-bromophenyl)hydrazine;hydrochloride
SMILES C1=CC(=CC=C1NN)Br.Cl

Specifications

Melting Point ∼215°C (decomposition)
Odor Odorless
Quantity 5 g
Beilstein 3565838
Solubility Information Soluble in water.
Formula Weight 223.51
Percent Purity 97%
Chemical Name or Material 4-Bromophenylhydrazine hydrochloride
Hazard Category H314
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 255-590-4
RTECSNumber MV0800000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only