missing translation for 'onlineSavingsMsg'
Learn More

4-Bromophenylacetylene, 97%

CAS: 766-96-1 | C8H5Br | 181.032 g/mol

Supplier:  Thermo Scientific Chemicals L1908203

 View more versions of this product

Catalog No. AAL1908203


Only null left
Explore available promotions

Description

Description

4-Bromophenylacetylene, is used as the starting material for second-order nonlinear optical materials, heterocyclotriynes, and unsymmetrical 1,4-diarylbutadiynes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromophenylacetylene, is used as the starting material for second-order nonlinear optical materials, heterocyclotriynes, and unsymmetrical 1,4-diarylbutadiynes.

Solubility
Soluble in chloroform, diethyl ether and ethanol.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
Specifications

Specifications

4-Bromophenylacetylene
64°C to 67°C
Odorless
MFCD00168822
1209726
Soluble in chloroform,diethyl ether and ethanol.
C#CC1=CC=C(C=C1)Br
181.032
181.04
766-96-1
88°C to 90°C (16 mmHg)
C8H5Br
1 g
4-bromophenylacetylene, benzene, 1-bromo-4-ethynyl, 1-bromo-4-ethynyl-benzene, 4-bromophenyl acetylene, 4'-bromophenyl acetylene, 4'-bromophenylacetylene, p-bromophenylacetylene, p-bromophenyl acetylene, 4-bromphenyl acetylene, 4-bromo-1-ethynylbenzene
LTLVZQZDXQWLHU-UHFFFAOYSA-N
1-bromo-4-ethynylbenzene
136603
97%

Chemical Identifiers

766-96-1
181.032
LTLVZQZDXQWLHU-UHFFFAOYSA-N
136603
C#CC1=CC=C(C=C1)Br
C8H5Br
MFCD00168822
4-bromophenylacetylene, benzene, 1-bromo-4-ethynyl, 1-bromo-4-ethynyl-benzene, 4-bromophenyl acetylene, 4'-bromophenyl acetylene, 4'-bromophenylacetylene, p-bromophenylacetylene, p-bromophenyl acetylene, 4-bromphenyl acetylene, 4-bromo-1-ethynylbenzene
1-bromo-4-ethynylbenzene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280i-P301+P312-P304+P340-P305+P351+P338-P312-P330-P501c

H302-H319-H335

DOTInformation : Hazard Class: 9; Packaging Group: III

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only