missing translation for 'onlineSavingsMsg'
Learn More

4-Bromophenylacetonitrile, 99%

Catalog No. AC154480250
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC154480250 25 g Glass bottle
AC154481000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC154480250 Supplier Thermo Scientific Chemicals Supplier No. 154480250

CAS: 16532-79-9 | C8H6BrN | 196.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 16532-79-9
Molecular Formula C8H6BrN
Molecular Weight (g/mol) 196.05
MDL Number MFCD00001916
InChI Key MFHFWRBXPQDZSA-UHFFFAOYSA-N
Synonym 4-bromophenylacetonitrile, 4-bromobenzyl cyanide, 2-4-bromophenyl acetonitrile, benzeneacetonitrile, 4-bromo, p-bromophenylacetonitrile, 4-bromobenzylcyanide, 4-bromobenzeneacetonitrile, p-bromobenzyl cyanide, p-bromobenzylcyanide, acetonitrile, p-bromophenyl
PubChem CID 27914
IUPAC Name 2-(4-bromophenyl)acetonitrile
SMILES BrC1=CC=C(CC#N)C=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 47°C to 49°C
Color Brown to Colorless
Flash Point 152°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula BrC6H4CH2CN
Quantity 25 g
Beilstein 09, 451
Solubility Information Solubility in water: insoluble
Formula Weight 196.05
Percent Purity 99%
Physical Form Crystalline Mass
Chemical Name or Material 4-Bromophenylacetonitrile, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if inhaled.
Toxic if swallowed.
Causes serious eye irritation.
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
If eye irr
EINECSNumber 240-602-2
RTECSNumber AL8090000
TSCA TSCA

RUO – Research Use Only