missing translation for 'onlineSavingsMsg'
Learn More

4-Bromophenylacetonitrile, 97%

Catalog No. AAA1088722
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1088722 100 g
AAA1088714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1088722 Supplier Thermo Scientific Chemicals Supplier No. A1088722
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 16532-79-9 | C8H6BrN | 196.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 16532-79-9
Molecular Formula C8H6BrN
Molecular Weight (g/mol) 196.05
MDL Number MFCD00001916
InChI Key MFHFWRBXPQDZSA-UHFFFAOYSA-N
Synonym 4-bromophenylacetonitrile, 4-bromobenzyl cyanide, 2-4-bromophenyl acetonitrile, benzeneacetonitrile, 4-bromo, p-bromophenylacetonitrile, 4-bromobenzylcyanide, 4-bromobenzeneacetonitrile, p-bromobenzyl cyanide, p-bromobenzylcyanide, acetonitrile, p-bromophenyl
PubChem CID 27914
IUPAC Name 2-(4-bromophenyl)acetonitrile
SMILES BrC1=CC=C(CC#N)C=C1

Specifications

Melting Point 45°C to 49°C
Density 1.49
Boiling Point 286°C
Flash Point 127°C (261°F)
Odor Pungent
Quantity 100 g
UN Number UN3449
Beilstein 1861069
Formula Weight 196.05
Percent Purity 97%
Chemical Name or Material 4-Bromophenylacetonitrile
Show More Show Less
Hazard Category H301-H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H311-H331-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 240-602-2
RTECSNumber AL8090000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only