missing translation for 'onlineSavingsMsg'
Learn More

4-Bromophenylacetic Acid 98.0+%, TCI America™

Catalog No. p-7118979
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B129625G 25 g
B1296250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B129625G Supplier TCI America Supplier No. B129625G
Only null left

Chemical Identifiers

CAS 1878-68-8
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.05
MDL Number MFCD00004342
InChI Key QOWSWEBLNVACCL-UHFFFAOYSA-N
Synonym 4-bromophenylacetic acid, 2-4-bromophenyl acetic acid, 4-bromophenyl acetic acid, p-bromophenylacetic acid, 4-bromobenzeneacetic acid, benzeneacetic acid, 4-bromo, p-bromophenyl acetic acid, acetic acid, p-bromophenyl, p-bromphenylessigsaure german
PubChem CID 74654
ChEBI CHEBI:1790
IUPAC Name 2-(4-bromophenyl)acetic acid
SMILES OC(=O)CC1=CC=C(Br)C=C1

Specifications

Melting Point 118°C
Color White-Yellow
Quantity 25 g
Formula Weight 215.05
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromophenylacetic Acid
RTECSNumber AF6325000
TSCA Yes