missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(4-Bromophenyl)urea 98.0+%, TCI America™
$63.38 - $63.38
Chemical Identifiers
| CAS | 1967-25-5 |
|---|---|
| Molecular Formula | C7H7BrN2O |
| Molecular Weight (g/mol) | 215.05 |
| MDL Number | MFCD00025428 |
| InChI Key | PFQUUCXMPUNRLA-UHFFFAOYSA-N |
| Synonym | 4-bromophenyl urea, 1-4-bromophenyl urea, n-4-bromophenyl urea, p-bromophenylurea, unii-n4k2d5wzsi, n4k2d5wzsi, urea, 4-bromophenyl, 4-bromophenyl-urea, acmc-209f0h, amino-n-4-bromophenyl amide |
| PubChem CID | 16074 |
| IUPAC Name | (4-bromophenyl)urea |
| SMILES | C1=CC(=CC=C1NC(=O)N)Br |
Chemical Identifiers
| 1967-25-5 | |
| 215.05 | |
| PFQUUCXMPUNRLA-UHFFFAOYSA-N | |
| 16074 | |
| C1=CC(=CC=C1NC(=O)N)Br |
| C7H7BrN2O | |
| MFCD00025428 | |
| 4-bromophenyl urea, 1-4-bromophenyl urea, n-4-bromophenyl urea, p-bromophenylurea, unii-n4k2d5wzsi, n4k2d5wzsi, urea, 4-bromophenyl, 4-bromophenyl-urea, acmc-209f0h, amino-n-4-bromophenyl amide | |
| (4-bromophenyl)urea |
Specifications
| 1967-25-5 | |
| Purple | |
| MFCD00025428 | |
| 4-bromophenyl urea, 1-4-bromophenyl urea, n-4-bromophenyl urea, p-bromophenylurea, unii-n4k2d5wzsi, n4k2d5wzsi, urea, 4-bromophenyl, 4-bromophenyl-urea, acmc-209f0h, amino-n-4-bromophenyl amide | |
| C1=CC(=CC=C1NC(=O)N)Br | |
| 215.05 | |
| 215.05 | |
| Crystalline Powder |
| 227°C | |
| C7H7BrN2O | |
| 5 g | |
| PFQUUCXMPUNRLA-UHFFFAOYSA-N | |
| (4-bromophenyl)urea | |
| 16074 | |
| ≥98.0% (N) | |
| (4-Bromophenyl)urea |
Safety and Handling
TSCA : No